Learn More
Cayman Chemical Estron 3-methyl ether 500mg
SDP
Catalog No. 502939305
Encompass_Preferred
A synthetic estrogen; stimulates proliferation of estrogen-dependent MCF-7 breast cancer cells in vitro from 0.16-20 μM; lacks ER binding activity (IC50s = >100 μM for ERα and ERβ); has been used in the synthesis of various estrogen receptor modulators
Catalog No. 50-293-9305 Supplier Cayman Chemical Supplier No. 27978500 MG
A synthetic estrogen; stimulates proliferation of estrogen-dependent MCF-7 breast cancer cells in vitro from 0.16-20 μM; lacks ER binding activity (IC50s = >100 μM for ERα and ERβ); has been used in the synthesis of various estrogen receptor modulators