Learn More
Cayman Chemical Estrone 3-methyl ether, Purity: ≥90%, CAS: 1624-62-0, 5 g
SDP
Catalog No. 502941435
Encompass_Preferred
A synthetic estrogen; stimulates proliferation of estrogen-dependent MCF-7 breast cancer cells in vitro from 0.16-20 μM; lacks ER binding activity (IC50s = >100 μM for ERα and ERβ); has been used in the synthesis of various estrogen receptor modulators
Catalog No. 50-294-1435 Supplier Cayman Chemical Supplier No. 279785 G
A synthetic estrogen; stimulates proliferation of estrogen-dependent MCF-7 breast cancer cells in vitro from 0.16-20 μM; lacks ER binding activity (IC50s = >100 μM for ERα and ERβ); has been used in the synthesis of various estrogen receptor modulators