Learn More
Cayman Chemical Estrone 3-sulfate (sodium salt), Purity: ≥95%, CAS: 438-67-5, 50 mg
SDP
Catalog No. 502915353
Encompass_Preferred
A steroid; selectively inhibits OATP1B1 over OATP1B3 (IC50s = 0.06 and 19.3 µM, respectively)
Catalog No. 50-291-5353 Supplier Cayman Chemical Supplier No. 3698950 MG
A steroid; selectively inhibits OATP1B1 over OATP1B3 (IC50s = 0.06 and 19.3 µM, respectively)