Learn More
Cayman Chemical Estron3GlucuronIdesodIum 10mg
SDP
Catalog No. 502946524
Encompass_Preferred
A metabolite of 17β-estradiol; urinary levels peak approximately 36 hours prior to ovulation and have been used as a marker of fertility in women
Catalog No. 50-294-6524 Supplier Cayman Chemical Supplier No. 3441710 MG
A metabolite of 17β-estradiol; urinary levels peak approximately 36 hours prior to ovulation and have been used as a marker of fertility in women