Learn More
Medchemexpress LLC Estrone-13C3 | 1241684-29-6 | 98.0% | 1 MG
SDP
Catalog No. 5000405439
Encompass_Preferred
Estrone-13C3 is the 13C-labeled version of Estrone, a naturally occurring estrogenic hormone. It is a primary endogenous estrogen, synthesized in various tissues through the aromatization of androstenedione. This compound is suitable for research use as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-05439 Supplier Medchemexpress LLC Supplier No. HYB0234S1MG
Estrone-13C3 is the 13C-labeled version of Estrone, a naturally occurring estrogenic hormone. It is a primary endogenous estrogen, synthesized in various tissues through the aromatization of androstenedione. This compound is suitable for research use as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS