Learn More
Medchemexpress LLC Estrone-16,16-d2 | 56588-58-0 | 95.0% | 272.38 g/mol | C18H20D2O2 | 10 MG
SDP
Catalog No. 5000298259
Encompass_Preferred
Estrone-d2-1 is a deuterium-labeled estrone provided for use as a stable isotope internal standard in analytical and mass spectrometry applications. The material is supplied as a 10 mg quantity with a reported purity of 95.0% (molecular formula C18H20D2O2, molecular weight 272.38 g/mol, CAS 56588-58-0).
  • Deuterium-labeled compound for use as an internal standard in quantitative analysis.
  • Suitable for mass spectrometry method development and validation.
  • Analytical-grade purity appropriate for research applications.
  • Compact 10 mg quantity convenient for laboratory use.
  • Supporting documentation available: datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-98259 Supplier Medchemexpress LLC Supplier No. HYB0234S410MG
Estrone-d2-1 is a deuterium-labeled estrone provided for use as a stable isotope internal standard in analytical and mass spectrometry applications. The material is supplied as a 10 mg quantity with a reported purity of 95.0% (molecular formula C18H20D2O2, molecular weight 272.38 g/mol, CAS 56588-58-0).
  • Deuterium-labeled compound for use as an internal standard in quantitative analysis.
  • Suitable for mass spectrometry method development and validation.
  • Analytical-grade purity appropriate for research applications.
  • Compact 10 mg quantity convenient for laboratory use.
  • Supporting documentation available: datasheet, certificate of analysis, and safety data sheet.