Learn More
Medchemexpress LLC Estrone 3-glucuronide sodium | 15087-01-1 | 98.3% | 468.47 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW0089171MG
Encompass_Preferred
Estrone 3-glucuronide sodium is a metabolite of 17-Estradiol. It is used as a standard to estimate estrone conjugate (E1Cs) from urine samples by enzyme immunoassays (EIAs) and is the reference standard used in assays that measure urinary E1C, a key marker for tracking the transition to the luteal phase.
  • Estimates estrone conjugate (E1Cs) from urine samples
  • Used in enzyme immunoassays (EIAs)
  • Reference standard in assays measuring urinary E1C
  • Key marker for tracking the transition to the luteal phase
  • Useful in urogenital, urologic, and endocrinology research
Catalog No. 50-004-53700