Learn More
Medchemexpress LLC Estrone 3-glucuronide sodium | 15087-01-1 | HY-W008917 | 98.8% | 468.47 | 5 MG
SDP
Catalog No. 5000451957
Encompass_Preferred
Estrone 3-glucuronide sodium is a metabolite of 17-Estradiol. It is used as a standard to estimate estrone conjugate (E1Cs) from urine samples by enzyme immunoassays (EIAs) and as a reference standard in assays that measure urinary E1C, a key marker for tracking the transition to the luteal phase.
  • Used as a standard to estimate estrone conjugate (E1Cs) from urine samples by enzyme immunoassays (EIAs).
  • Reference standard in assays that measure urinary E1C.
  • Key marker for tracking the transition to the luteal phase.
Catalog No. 50-004-51957 Supplier Medchemexpress LLC Supplier No. HYW0089175MG
Add to Cart
Add to Cart
Estrone 3-glucuronide sodium is a metabolite of 17-Estradiol. It is used as a standard to estimate estrone conjugate (E1Cs) from urine samples by enzyme immunoassays (EIAs) and as a reference standard in assays that measure urinary E1C, a key marker for tracking the transition to the luteal phase.
  • Used as a standard to estimate estrone conjugate (E1Cs) from urine samples by enzyme immunoassays (EIAs).
  • Reference standard in assays that measure urinary E1C.
  • Key marker for tracking the transition to the luteal phase.