Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estrone 3-methyl ether | 1624-62-0 | 97.38% | C19H24O2 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY795765MG

Encompass_Preferred

Estrone 3-methyl ether is a synthetic intermediate used in the synthesis of estrogen receptor modulators.

  • Used in the synthesis of estrogen receptor modulators
  • Molecular weight: 284.39
  • Molecular formula: C19H24O2
  • CAS number: 1624-62-0
  • Purity: 97.38%
  • Appearance: Solid, white to off-white
  • Storage (Powder): -20°C for 3 years; 4°C for 2 years
  • Storage (In solvent): -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-32845


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.