Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estrone 3-methyl ether | 1624-62-0 | MFCD00046228 | 97.4% | 284.39 g/mol | C19H24O2 | 10 MG
SDP

Catalog No. 5000298775
Encompass_Preferred

Estrone 3-methyl ether is a synthetic steroid intermediate used in organic synthesis and research on estrogen receptor modulators. It is supplied as a solid with high purity and is used as a building block in medicinal chemistry and assay development.

  • CAS number: 1624-62-0.
  • Molecular formula: C19H24O2.
  • Molecular weight: 284.39 g/mol.
  • Purity: 97.4%.
  • Physical form: solid.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Common applications: synthesis of estrogen receptor modulators and medicinal chemistry building block.

Catalog No. 50-002-98775 Supplier Medchemexpress LLC Supplier No. HY7957610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Estrone 3-methyl ether is a synthetic steroid intermediate used in organic synthesis and research on estrogen receptor modulators. It is supplied as a solid with high purity and is used as a building block in medicinal chemistry and assay development.

  • CAS number: 1624-62-0.
  • Molecular formula: C19H24O2.
  • Molecular weight: 284.39 g/mol.
  • Purity: 97.4%.
  • Physical form: solid.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (6 months) or -20°C (1 month).
  • Common applications: synthesis of estrogen receptor modulators and medicinal chemistry building block.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.