Learn More
Alpco Ltd Equine Estrone-3-sulfate ELISA
SDP
Supplier:  Alpco Ltd 55E3SEQE01
Encompass_Preferred
Equine Estrone-3-sulfate ELISA is for the quantitative determination of estrone-3-sulfate in mare serum. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-751-3633