Learn More
Apexbio Technology LLC Estrone 53-16-7 10g
SDP
Supplier:  Apexbio Technology LLC A842610000
Encompass_Preferred
Estrone (53-16-7) is a naturally occurring estrogen that modulates estrogen receptor-mediated signaling pathways It is primarily synthesized in ovarian and adipose tissues and participates in the regulation of estrogen-dependent cellular and physiological processes Estrone exerts its biological activity primarily through binding and activating estrogen receptors Experimental model activity type and key metric (IC50/EC50) are not specified Based on these pharmacological properties estrone holds research potential in studies of estrogen receptor activation reproductive endocrinology menopausal hormone transitions and estrogen-dependent signaling pathways and diseases
Catalog No. 50-000-93859