Learn More
Medchemexpress LLC Estrone | 53-16-7 | 99.9% | 5 G

Supplier: Medchemexpress LLC HYB02345G
Estrone (E1) is a natural estrogenic hormone and the primary endogenous estrogen in postmenopausal women. It is produced by several tissues, particularly adipose tissue, through the aromatization of androstenedione.
- Natural estrogenic hormone and endogenous estrogen
- Produced in adipose tissue
- Targets estrogen receptor and ERR
- Relevant for endocrinology and cancer research
- Categorized as a natural product, inhibitor, or agonist
- Involved in vitamin D related and nuclear receptor pathways
- Involved in metabolic enzyme and protease pathways
- Stable as powder at -20°C for 3 years and 4°C for 2 years
- Stable in solvent at -80°C for 6 months and -20°C for 1 month
- Soluble in DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.