Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estrone | 53-16-7 | 99.96% | 1 ML
SDP

Catalog No. 5000405438
Encompass_Preferred

Estrone (E1) is a natural estrogenic hormone and the main representative of endogenous estrogens. It is produced by several tissues, particularly adipose tissue, through the aromatization of androstenedione in fat cells.

  • Purity of 99.96%
  • Molecular weight of 270.37
  • Chemical formula C18H22O2
  • Appears as a solid, white to off-white
  • Initial source from microorganisms or as an endogenous metabolite
  • Available as a 10 mM, 1 mL solution in DMSO
  • Soluble in DMSO (25 mg/mL or 92.47 mM)
  • Insoluble in water
  • For research use only

Catalog No. 50-004-05438 Supplier Medchemexpress LLC Supplier No. HYB023410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Estrone (E1) is a natural estrogenic hormone and the main representative of endogenous estrogens. It is produced by several tissues, particularly adipose tissue, through the aromatization of androstenedione in fat cells.

  • Purity of 99.96%
  • Molecular weight of 270.37
  • Chemical formula C18H22O2
  • Appears as a solid, white to off-white
  • Initial source from microorganisms or as an endogenous metabolite
  • Available as a 10 mM, 1 mL solution in DMSO
  • Soluble in DMSO (25 mg/mL or 92.47 mM)
  • Insoluble in water
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.