Learn More
Medchemexpress LLC Estrone-d2-1 | 56588-58-0 | 99.8% | 1 MG
SDP
Catalog No. 5000406126
Encompass_Preferred
Estrone-d2-1 is a deuterium-labeled form of Estrone (E1), a natural estrogenic hormone produced in various tissues like adipose tissue. It is formed through the aromatization of androstenedione in fat cells. This product is for research use only.
  • Stable heavy isotopes incorporated into drug molecules for use as tracers.
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High purity: 99.75%.
Catalog No. 50-004-06126 Supplier Medchemexpress LLC Supplier No. HYB0234S41MG
Estrone-d2-1 is a deuterium-labeled form of Estrone (E1), a natural estrogenic hormone produced in various tissues like adipose tissue. It is formed through the aromatization of androstenedione in fat cells. This product is for research use only.
  • Stable heavy isotopes incorporated into drug molecules for use as tracers.
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • High purity: 99.75%.