Learn More
Medchemexpress LLC Estrone-d2 | 350820-16-5 | 99.78% | 1 MG
SDP
Catalog No. 5000405695
Encompass_Preferred
Estrone-d2 is a deuterium-labeled form of Estrone (E1), a natural estrogenic hormone primarily produced by various tissues. It is intended for research use only and serves as a tracer or internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Used as a tracer for quantitative analysis
  • Functions as an internal standard for quantitative analysis
  • Deuterium-labeled form of estrone
  • For research use only
Catalog No. 50-004-05695 Supplier Medchemexpress LLC Supplier No. HYB0234S11MG
Estrone-d2 is a deuterium-labeled form of Estrone (E1), a natural estrogenic hormone primarily produced by various tissues. It is intended for research use only and serves as a tracer or internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
  • Used as a tracer for quantitative analysis
  • Functions as an internal standard for quantitative analysis
  • Deuterium-labeled form of estrone
  • For research use only