Learn More
Medchemexpress LLC Estrone-d4 | 53866-34-5 | 99.5% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0234S21MG
Encompass_Preferred
Estrone-d4 is the deuterium-labeled version of Estrone (E1), a natural estrogenic hormone. Estrone is a primary endogenous estrogen, produced predominantly by adipose tissue. It is formed through the aromatization of androstenedione in fat cells. Deuterium substitution in drug molecules is used as a tracer for quantitation in drug development and has the potential to influence pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06369