Learn More
Biomatik Corporation Estrone (E1) CLIA Kit
Supplier:  Biomatik Corporation EKU08652
Encompass_Preferred
Estrone (E1) CLIA Kit for detecting E1 in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 23.44-6000pg/ml. Sensitivity: 7.96pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Target Synonyms: Oestrone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6448