Learn More
Biomatik Corporation Estrone (E1) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11102
Encompass_Preferred
Estrone (E1) Mini sample ELISA Kit for detecting E1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 24.69-2,000pg/ml. Sensitivity: 8.83pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: Oestrone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8224