Learn More
Medchemexpress LLC Estrone O-sulfamate | 148672-09-7 | 99.99% | C18H23NO4S | 1 MG
SDP
Supplier:  Medchemexpress LLC HY145851MG
Encompass_Preferred
Estrone O-sulfamate (Estrone 3-O-sulfamate) is a potent steroid sulfatase (STS) inhibitor. It can be used for cancer research.
  • Potent steroid sulfatase (STS) inhibitor
  • Demonstrates inhibitory activity in placental microsomes (P.M.) and MCF-7 cells
  • Applicable for cancer research
Catalog No. 50-003-56127