Learn More
Medchemexpress LLC Estrone O-sulfamate | 148672-09-7 | 99.99% | C18H23NO4S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY14585100MG
Encompass_Preferred
Estrone O-sulfamate (Estrone 3-O-sulfamate) is a potent steroid sulfatase (STS) inhibitor. It has inhibitory activity for STS in a placental microsomes (P.M.) preparation and in MCF-7 cells with IC50 values of 18 nM and 0.83 nM, respectively. This compound can be used for the research of cancer.
  • Potent steroid sulfatase (STS) inhibitor.
  • Inhibitory activity in placental microsomes (P.M.) preparation with an IC50 of 18 nM.
  • Inhibitory activity in MCF-7 cells with an IC50 of 0.83 nM.
  • Suitable for cancer research.
Catalog No. 50-003-54045