Learn More
Medchemexpress LLC Estrone (Standard) | 53-16-7 | 99.7% | 25 MG
SDP
Catalog No. 5000450649
Encompass_Preferred
Estrone (Standard) is the analytical standard of Estrone and is intended for research and analytical applications. Estrone (E1) is a natural estrogenic hormone, representing a main endogenous estrogen produced by various tissues, particularly adipose tissue. It is formed through the aromatization of androstenedione in fat cells.
  • Used as an analytical standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Microorganisms, endogenous metabolite
  • Store under recommended conditions in the Certificate of Analysis
Catalog No. 50-004-50649 Supplier Medchemexpress LLC Supplier No. 25MGHYB0234R
Estrone (Standard) is the analytical standard of Estrone and is intended for research and analytical applications. Estrone (E1) is a natural estrogenic hormone, representing a main endogenous estrogen produced by various tissues, particularly adipose tissue. It is formed through the aromatization of androstenedione in fat cells.
  • Used as an analytical standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS
  • Appearance: Solid
  • Color: White to off-white
  • Initial source: Microorganisms, endogenous metabolite
  • Store under recommended conditions in the Certificate of Analysis