Learn More
Cayman Chemical Estrone, Purity: ≥95%, CAS: 53-16-7, 1 g
SDP
Catalog No. 502888245
Encompass_Preferred
A primary, naturally-occurring estrogen; serum concentrations in premenopausal women fluctuate according to the menstrual cycle; becomes the most predominant estrogen in postmenopausal women; binding affinities to ERα and ERβ are ~60% and ~37% relative to estradiol
Catalog No. 50-288-8245 Supplier Cayman Chemical Supplier No. 100064851 G
A primary, naturally-occurring estrogen; serum concentrations in premenopausal women fluctuate according to the menstrual cycle; becomes the most predominant estrogen in postmenopausal women; binding affinities to ERα and ERβ are ~60% and ~37% relative to estradiol