Learn More
Cayman Chemical Estrone, Purity: ≥95%, CAS: 53-16-7, 500 mg
SDP
Catalog No. 502890466
Encompass_Preferred
A primary, naturally-occurring estrogen; serum concentrations in premenopausal women fluctuate according to the menstrual cycle; becomes the most predominant estrogen in postmenopausal women; binding affinities to ERα and ERβ are ~60% and ~37% relative to estradiol
Catalog No. 50-289-0466 Supplier Cayman Chemical Supplier No. 10006485500 MG
A primary, naturally-occurring estrogen; serum concentrations in premenopausal women fluctuate according to the menstrual cycle; becomes the most predominant estrogen in postmenopausal women; binding affinities to ERα and ERβ are ~60% and ~37% relative to estradiol