Learn More
Cayman Chemical Estrone Steroid Hormones
SDP
Supplier:  Cayman Chemical 1000648510 G
Encompass_Preferred
A primary, naturally-occurring estrogen; serum concentrations in premenopausal women fluctuate according to the menstrual cycle; becomes the most predominant estrogen in postmenopausal women; binding affinities to ERα and ERβ are ~60% and ~37% relative to estradiol
Catalog No. 50-289-1314