Learn More
Cayman Chemical Estrone Steroid Hormones
SDP
Catalog No. 502888244
Encompass_Preferred
A primary, naturally-occurring estrogen; serum concentrations in premenopausal women fluctuate according to the menstrual cycle; becomes the most predominant estrogen in postmenopausal women; binding affinities to ERα and ERβ are ~60% and ~37% relative to estradiol
Catalog No. 50-288-8244 Supplier Cayman Chemical Supplier No. 100064855 G
Add to Cart
Add to Cart
A primary, naturally-occurring estrogen; serum concentrations in premenopausal women fluctuate according to the menstrual cycle; becomes the most predominant estrogen in postmenopausal women; binding affinities to ERα and ERβ are ~60% and ~37% relative to estradiol