Learn More
Medchemexpress LLC Estrone sulfate-d4 sodium | 285979-80-8 | MFCD01075457 | 99.4% | 376.44 g/mol | C18H17D4NaO5S | 5 MG
SDP
Catalog No. 5000264795
Encompass_Preferred
Estrone sulfate-d4 sodium is a deuterium-labeled (stable isotope) sodium salt of estrone sulfate supplied as an analytical reference standard. It is intended as an internal standard for LC-MS/MS and other mass spectrometry applications, and for pharmacokinetic and metabolic research.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (99.38%).
  • White to off-white solid suitable for analytical handling.
  • Available in 1 mg, 5 mg, 10 mg, and 50 mg packages.
  • Recommended storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-64795 Supplier Medchemexpress LLC Supplier No. HY113293BS15MG
Estrone sulfate-d4 sodium is a deuterium-labeled (stable isotope) sodium salt of estrone sulfate supplied as an analytical reference standard. It is intended as an internal standard for LC-MS/MS and other mass spectrometry applications, and for pharmacokinetic and metabolic research.
  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (99.38%).
  • White to off-white solid suitable for analytical handling.
  • Available in 1 mg, 5 mg, 10 mg, and 50 mg packages.
  • Recommended storage: 4°C sealed; in solvent: -80°C (6 months), -20°C (1 month).