Learn More
Medchemexpress LLC Estrone sulfate (potassium) | 1240-04-6 | 99.9% | 388.52 | 1 ML
SDP
Catalog No. 5000437692
Encompass_Preferred
Estrone sulfate potassium is an inactive endogenous estrogen that can be converted into Estrone and Estradiol. It is also a substrate of the OATP1B3 transporter. This compound is applicable to breast cancer-related research, as it stimulates the growth of nitrosomethylurea-induced mammary tumors and promotes colony formation of dispersed nitrosomethylurea mammary cells through its conversion to Estrone and Estradiol.
  • Taken up at a low rate by normal breast parenchymal cells.
  • Converted to estrone by normal breast parenchymal cells.
  • Stimulates colony formation in dispersed nitrosomethylurea mammary cells.
  • Exerts dose-related stimulatory effects on nitrosomethylurea-induced hormone-responsive mammary tumor growth in castrate rats.
  • Reduces glucose-6-phosphatase activity and protein levels, and lowers blood glucose in ob/ob mice.
  • Modulates glucose-6-phosphatase kinetic parameters in control mice.
Catalog No. 50-004-37692 Supplier Medchemexpress LLC Supplier No. HY113293A10MM/1ML
Estrone sulfate potassium is an inactive endogenous estrogen that can be converted into Estrone and Estradiol. It is also a substrate of the OATP1B3 transporter. This compound is applicable to breast cancer-related research, as it stimulates the growth of nitrosomethylurea-induced mammary tumors and promotes colony formation of dispersed nitrosomethylurea mammary cells through its conversion to Estrone and Estradiol.
  • Taken up at a low rate by normal breast parenchymal cells.
  • Converted to estrone by normal breast parenchymal cells.
  • Stimulates colony formation in dispersed nitrosomethylurea mammary cells.
  • Exerts dose-related stimulatory effects on nitrosomethylurea-induced hormone-responsive mammary tumor growth in castrate rats.
  • Reduces glucose-6-phosphatase activity and protein levels, and lowers blood glucose in ob/ob mice.
  • Modulates glucose-6-phosphatase kinetic parameters in control mice.