Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Estrone sulfate potassium | 1240-04-6 | MFCD00056410 | 99.5% | 388.52 | C18H21KO5S | 5 MG
SDP

Catalog No. 5000263543
Encompass_Preferred

Estrone sulfate potassium is the potassium salt of estrone sulfate, an endogenous estrogen conjugate supplied for laboratory research. It is available as a high-purity solid and as a DMSO solution for in vitro studies, and is intended for research use only.

  • High purity 99.45%.
  • Molecular weight 388.52 and formula C18H21KO5S.
  • Available in multiple sizes, including 5 mg, and as a 10 mM solution in DMSO.
  • Soluble in DMSO for preparation of stock solutions.
  • Supplied for research use only; not for human or clinical use.
  • May be restricted or controlled in some territories.

Catalog No. 50-002-63543 Supplier Medchemexpress LLC Supplier No. HY113293A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Estrone sulfate potassium is the potassium salt of estrone sulfate, an endogenous estrogen conjugate supplied for laboratory research. It is available as a high-purity solid and as a DMSO solution for in vitro studies, and is intended for research use only.

  • High purity 99.45%.
  • Molecular weight 388.52 and formula C18H21KO5S.
  • Available in multiple sizes, including 5 mg, and as a 10 mM solution in DMSO.
  • Soluble in DMSO for preparation of stock solutions.
  • Supplied for research use only; not for human or clinical use.
  • May be restricted or controlled in some territories.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.