Learn More
TARGETMOL CHEMICALS INC Estrone sulfate sodium 25MG
SDP
Catalog No. 502878862
Encompass_Preferred
Also available in 2 mg, 5 mg, 10 mg, 50 mg and bulk. Please contact Fisher for quotes. Estrone sulfate sodium (17beta-Estrone 3-sulfate) is an endogenous steroid and an estrogen ester that is biologically inactive. It is converted by steroid sulfatase into estrone . Estrone sulfate sodium has been investigated as a ligand for targeting organic anion transporting polypeptides f Purity 98%
Catalog No. 50-287-8862 Supplier TARGETMOL CHEMICALS INC Supplier No. T3685725MG
Also available in 2 mg, 5 mg, 10 mg, 50 mg and bulk. Please contact Fisher for quotes. Estrone sulfate sodium (17beta-Estrone 3-sulfate) is an endogenous steroid and an estrogen ester that is biologically inactive. It is converted by steroid sulfatase into estrone . Estrone sulfate sodium has been investigated as a ligand for targeting organic anion transporting polypeptides f Purity 98%