Learn More
Medchemexpress LLC Estrone sulfate sodium | 438-67-5 | 99.8% | 372.41 | 1 ML
SDP
Catalog No. 5000438626
Encompass_Preferred
Estrone sulfate sodium is an inactive endogenous estrogen that can be converted into Estrone and Estradiol. It is a substrate of the OATP1B3 transporter and is applicable to breast cancer-related research. It stimulates the growth of nitrosomethylurea-induced mammary tumors and colony formation of dispersed nitrosomethylurea mammary cells.
  • Inactive endogenous estrogen
  • Converted to estrone and estradiol
  • Substrate of OATP1B3 transporter
  • Stimulates mammary tumor growth
  • Applicable to breast cancer research
Catalog No. 50-004-38626 Supplier Medchemexpress LLC Supplier No. HY113293B10MM/1ML
Estrone sulfate sodium is an inactive endogenous estrogen that can be converted into Estrone and Estradiol. It is a substrate of the OATP1B3 transporter and is applicable to breast cancer-related research. It stimulates the growth of nitrosomethylurea-induced mammary tumors and colony formation of dispersed nitrosomethylurea mammary cells.
  • Inactive endogenous estrogen
  • Converted to estrone and estradiol
  • Substrate of OATP1B3 transporter
  • Stimulates mammary tumor growth
  • Applicable to breast cancer research