Learn More
Medchemexpress LLC Estrone sulfate (sodium) | 438-67-5 | 99.8% | 372.41 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY113293B
Encompass_Preferred
Estrone sulfate sodium is an inactive endogenous estrogen. It can be converted into Estrone and Estradiol and is a substrate of the OATP1B3 transporter. This compound stimulates the growth of nitrosomethylurea-induced mammary tumors and colony formation of dispersed nitrosomethylurea mammary cells, with conversion to Estrone and Estradiol occurring both in vivo and in vitro.
  • Can be converted into Estrone and Estradiol.
  • Substrate of the OATP1B3 transporter.
  • Stimulates mammary tumor growth in ovariectomized rats.
  • Applicable to breast cancer-related research.
Catalog No. 50-004-39280