Learn More
Cayman Chemical Estropipate, Purity: ≥98%, CAS: 7280-37-7, 10 mg
SDP
Catalog No. 502995107
Encompass_Preferred
A piperazine salt form of estrone 3-sulfate; inhibits OATP1B1 selectively over OATP1B3 (IC50s = 0.06 and 19.32 µM, respectively)
Catalog No. 50-299-5107 Supplier Cayman Chemical Supplier No. 2380610 MG
A piperazine salt form of estrone 3-sulfate; inhibits OATP1B1 selectively over OATP1B3 (IC50s = 0.06 and 19.32 µM, respectively)