Learn More
TARGETMOL CHEMICALS INC ESUPRONE 5MG
SDP
Catalog No. 502580631
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Esuprone (LU-43839) is a novel reversible and highly selective MAO-A inhibitor with anticonvulsant activity for the treatment of depression. purity: 99%
Catalog No. 50-258-0631 Supplier TARGETMOL CHEMICALS INC Supplier No. T272885MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Esuprone (LU-43839) is a novel reversible and highly selective MAO-A inhibitor with anticonvulsant activity for the treatment of depression. purity: 99%