Learn More
Aviva Systems Biology Corp ESX1 Antibody - N-terminal region (ARP36848_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36848_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ESX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse ESX1. Alternate names: Spx1ApplicationWB. Species reactivity: Mouse
Catalog No. 50-014-53059