Learn More
Aviva Systems Biology Corp ESX1 Antibody - N-terminal region (ARP36220_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36220_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ESX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ESX1. Alternate names: ESX1L, ESXR1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53039