Learn More
ABclonal Technology ESYT1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A154101000UL
Encompass_Preferred
Enables identical protein binding activity. Predicted to be involved in endoplasmic reticulum-plasma membrane tethering and lipid transport. Located in endoplasmic reticulum. Is integral component of endoplasmic reticulum membrane.
Catalog No. 50-001-08032