Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Et-29 | 2166487-23-4 | 99.9% | C34H46N6O6S | 1 ML
SDP

Catalog No. 5000356406
Encompass_Preferred

Et-29 is a potent and selective SIRT5 inhibitor, primarily intended for research use. It exhibits a Ki of 40 nM.

  • Potent and selective SIRT5 inhibitor
  • For research use only
  • White to off-white solid appearance
  • Molecular weight: 666.83
  • Purity: 99.9%
  • Storage as solid: -20°C, protect from light, under nitrogen
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month, protect from light, under nitrogen

Catalog No. 50-003-56406 Supplier Medchemexpress LLC Supplier No. HY14565110MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Et-29 is a potent and selective SIRT5 inhibitor, primarily intended for research use. It exhibits a Ki of 40 nM.

  • Potent and selective SIRT5 inhibitor
  • For research use only
  • White to off-white solid appearance
  • Molecular weight: 666.83
  • Purity: 99.9%
  • Storage as solid: -20°C, protect from light, under nitrogen
  • Storage in solvent: -80°C for 6 months; -20°C for 1 month, protect from light, under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.