Learn More
Cole-Parmer Thermo Scientific Nalgene 5000-1012 Cryogenic vial; 1.0 mL, 25/pack
Catalog No. NC0438385 Shop All Cole-Parmer Products
Encompass_Preferred
White marking area aids in sample identification. Graduated on the side. Order colored cap inserts separately. Note: Do not autoclave or immerse vials in liquid nitrogen. For storage under extreme conditions such as liquid nitrogen, use CryoFlex™ tubing 03755-64 (order separately).
Catalog No. NC0438385 Supplier Cole-Parmer Supplier No. 0675493
White marking area aids in sample identification. Graduated on the side. Order colored cap inserts separately. Note: Do not autoclave or immerse vials in liquid nitrogen. For storage under extreme conditions such as liquid nitrogen, use CryoFlex™ tubing 03755-64 (order separately).