Learn More
New England Biolabs, Inc. ET SSB – 50 µg
SDP
Supplier:  New England Biolabs, Inc. M2401S
Encompass
ET SSB (Extreme Thermostable Single-Stranded DNA Binding Protein) is a single-stranded DNA binding protein isolated from a hyperthermophilic microorganism, which remains fully active after incubation at 95C for 60 minutes. Due to the extreme thermostability, ET SSB can be used in applications that require extremely high temperature conditions, such as nucleic acid amplification and sequencing.
Catalog No. 50-994-967