Learn More
Apexbio Technology LLC ET SSB Protein 100ug
SDP
Catalog No. 5000096355
Encompass_Preferred
ET SSB Protein is a recombinant protein that selectively binds to single-stranded DNA (ssDNA) It is designed to stabilize unfolded DNA regions and prevent strand reassociation or nuclease degradation ET SSB Protein exerts its biological activity primarily by binding ssDNA maintaining DNA in a single-stranded state and withstanding incubation at temperatures up to 95 C Based on these properties ET SSB Protein holds research potential in PCR and q-PCR assay optimization enhancement of DNA polymerase performance stabilization and labeling of ssDNA facilitation of reverse transcriptase efficiency in RT-PCR and sequencing protocols involving strong secondary structures
Catalog No. 50-000-96355 Supplier Apexbio Technology LLC Supplier No. P20080.1
ET SSB Protein is a recombinant protein that selectively binds to single-stranded DNA (ssDNA) It is designed to stabilize unfolded DNA regions and prevent strand reassociation or nuclease degradation ET SSB Protein exerts its biological activity primarily by binding ssDNA maintaining DNA in a single-stranded state and withstanding incubation at temperatures up to 95 C Based on these properties ET SSB Protein holds research potential in PCR and q-PCR assay optimization enhancement of DNA polymerase performance stabilization and labeling of ssDNA facilitation of reverse transcriptase efficiency in RT-PCR and sequencing protocols involving strong secondary structures