Learn More
Enzo Life Sciences Endothelin-1 (human, mouse, rat, pig, bovine, dog) (1mg). CAS: 117399-94-7
SDP
Supplier:  Enzo Life Sciences ALX155001P001
Encompass_Preferred
Alternative name: ET-1. Formula: C109H159N25O32S5. MW: 2492.0. Purity: ≥98% (HPLC). Formulation: Lyophilized. MI: 14: 3576. Solubility: 1mg/ml soluble in DMSO or 5% acetic acid. Long Term Storage: -20°C. Use/Stability: Solution is stable for 1-2 weeks when stored at -20°C. Prepare solution just before use.
Catalog No. 50-200-7791