Learn More
Enzo Life Sciences Endothelin A receptor polyclonal antibody (250µg)
SDP
Supplier:  Enzo Life Sciences ALX210507AC250
Encompass_Preferred
Host: Sheep. Immunogen: Synthetic peptide corresponding to aa 410-422 (Q410EQNHNTERSSHK422) of C-terminal rat ETA-receptor (endothelin A receptor). Species reactivity: Human, Mouse, Rat. Specificity: Recognizes an epitope on the cytoplasmic side of the ETA-receptor. Applications: IHC, WB. Recommended Dilutions: Immunohistochemistry (paraffin sections, 4µg/ml) Western Blot (10-20µg/ml using ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Epitope-affinity purified. Formulation: Liquid. In TRIS-citrate-phosphate, pH 7-8, containing 0.1% sodium azide. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -20°C.
Catalog No. 50-200-8125