Learn More
TARGETMOL CHEMICALS INC ETAROTENE 5MG
SDP
Catalog No. 502580754
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Etarotene (Arotinoid ethyl sulphone) is an ethylsulfonyl derivative of aloe acid with differentiation-inducing and potentially antitumor activity.Etarotene is a collagenase inhibitor and can be used to study psoriasis. purity: 100%
Catalog No. 50-258-0754 Supplier TARGETMOL CHEMICALS INC Supplier No. T253935MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Etarotene (Arotinoid ethyl sulphone) is an ethylsulfonyl derivative of aloe acid with differentiation-inducing and potentially antitumor activity.Etarotene is a collagenase inhibitor and can be used to study psoriasis. purity: 100%