Learn More
Medchemexpress LLC Etazolate hydrochloride | 35838-58-5 | MFCD00209848 | 98.0% | 325.79 | C14H20ClN5O2 | 5 MG
SDP
Catalog No. 5000212249
Encompass_Preferred
Etazolate hydrochloride is a pyrazolopyridine research compound that functions as a selective phosphodiesterase-4 (PDE4) inhibitor, a GABAA receptor regulator, and an α-secretase activator. It is used in neuropharmacology and inflammation studies to probe cAMP signaling, amyloid precursor protein processing, and behavioral pharmacology.
Catalog No. 50-002-12249 Supplier Medchemexpress LLC Supplier No. HY1009365MG
Etazolate hydrochloride is a pyrazolopyridine research compound that functions as a selective phosphodiesterase-4 (PDE4) inhibitor, a GABAA receptor regulator, and an α-secretase activator. It is used in neuropharmacology and inflammation studies to probe cAMP signaling, amyloid precursor protein processing, and behavioral pharmacology.