Learn More
Medchemexpress LLC Etazolate hydrochloride | 35838-58-5 | MFCD00209848 | 98.0% | 325.79 | C14H20ClN5O2 | 50 MG
SDP
Catalog No. 5000212419
Encompass_Preferred
Etazolate hydrochloride is the hydrochloride salt of etazolate, a pyrazolopyridine-derived small molecule used in research as a selective phosphodiesterase-4 (PDE4) inhibitor and a GABAA receptor modulator. It is provided as a high-purity research reagent intended for in vitro and in vivo studies investigating neurological function, inflammation, and related signaling pathways.
  • High-purity hydrochloride salt suitable for research use.
  • Selective phosphodiesterase-4 (PDE4) inhibition activity.
  • Modulates GABAA receptor function.
  • Supplied in a convenient small-scale pack for laboratory studies.
  • Characterized by molecular formula and molecular weight for traceability.
Catalog No. 50-002-12419 Supplier Medchemexpress LLC Supplier No. HY10093650MG
Etazolate hydrochloride is the hydrochloride salt of etazolate, a pyrazolopyridine-derived small molecule used in research as a selective phosphodiesterase-4 (PDE4) inhibitor and a GABAA receptor modulator. It is provided as a high-purity research reagent intended for in vitro and in vivo studies investigating neurological function, inflammation, and related signaling pathways.
  • High-purity hydrochloride salt suitable for research use.
  • Selective phosphodiesterase-4 (PDE4) inhibition activity.
  • Modulates GABAA receptor function.
  • Supplied in a convenient small-scale pack for laboratory studies.
  • Characterized by molecular formula and molecular weight for traceability.