Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Etazolate hydrochloride | 35838-58-5 | MFCD00209848 | 98.0% | 325.79 g/mol | C14H20ClN5O2 | 1 ML
SDP

Catalog No. 5000212297
Encompass_Preferred

Etazolate hydrochloride is a research-grade small molecule inhibitor of phosphodiesterase 4 (PDE4) and a modulator of GABAA receptors. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) or as a solid for reconstitution, and is used in neuroscience and pharmacology studies investigating neuroprotection, inflammation, and amyloid precursor protein processing.

  • Selective PDE4 inhibitor and GABAA receptor modulator.
  • Supplied as a 10 mM solution in DMSO (1 mL) or as solid for reconstitution.
  • Purity ≥98.0% as reported by supplier.
  • Molecular weight approximately 325.8 g/mol.
  • CAS number 35838-58-5 for unambiguous identification.
  • Available in small milligram quantities suitable for research use.
  • Intended for research applications; not for human or clinical use.

Catalog No. 50-002-12297 Supplier Medchemexpress LLC Supplier No. HY10093610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Etazolate hydrochloride is a research-grade small molecule inhibitor of phosphodiesterase 4 (PDE4) and a modulator of GABAA receptors. It is provided as a ready-to-use 10 mM solution in DMSO (1 mL) or as a solid for reconstitution, and is used in neuroscience and pharmacology studies investigating neuroprotection, inflammation, and amyloid precursor protein processing.

  • Selective PDE4 inhibitor and GABAA receptor modulator.
  • Supplied as a 10 mM solution in DMSO (1 mL) or as solid for reconstitution.
  • Purity ≥98.0% as reported by supplier.
  • Molecular weight approximately 325.8 g/mol.
  • CAS number 35838-58-5 for unambiguous identification.
  • Available in small milligram quantities suitable for research use.
  • Intended for research applications; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.