Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Etazolate hydrochloride | 35838-58-5 | MFCD00209848 | 98.0% | 325.79 g/mol | C14H20ClN5O2 | 100 MG
SDP

Catalog No. 5000212615
Encompass_Preferred

Etazolate hydrochloride is the hydrochloride salt of etazolate, a research compound that selectively inhibits phosphodiesterase-4 (PDE4) and modulates GABAA receptors. It is provided as a solid for preclinical biochemical and neuroscience research, including studies of inflammation and neuroprotection.

  • Selective PDE4 inhibitor and GABAA receptor modulator.
  • Hydrochloride salt form, solid suitable for laboratory use.
  • Purity approximately 98.0%.
  • Molecular weight 325.79 g/mol.
  • CAS number 35838-58-5.
  • Supplied in a 100 mg pack for bench-scale experiments.

Catalog No. 50-002-12615 Supplier Medchemexpress LLC Supplier No. HY100936100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Etazolate hydrochloride is the hydrochloride salt of etazolate, a research compound that selectively inhibits phosphodiesterase-4 (PDE4) and modulates GABAA receptors. It is provided as a solid for preclinical biochemical and neuroscience research, including studies of inflammation and neuroprotection.

  • Selective PDE4 inhibitor and GABAA receptor modulator.
  • Hydrochloride salt form, solid suitable for laboratory use.
  • Purity approximately 98.0%.
  • Molecular weight 325.79 g/mol.
  • CAS number 35838-58-5.
  • Supplied in a 100 mg pack for bench-scale experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.