Learn More
Medchemexpress LLC Etazolate hydrochloride | 35838-58-5 | MFCD00209848 | 98.0% | 325.79 g·mol⁻¹ | C14H20ClN5O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10093625MG
Encompass_Preferred
Etazolate hydrochloride is a small-molecule phosphodiesterase-4 (PDE4) inhibitor and GABAA receptor modulator supplied as a hydrochloride salt for research use. It is typically provided as a solid with high purity and is used in pharmacology and neuroscience studies to probe PDE4-mediated signaling and neuroprotective or antidepressant-like effects.
  • High purity (98.0%).
  • Molecular formula C14H20ClN5O2 and molecular weight 325.79 g·mol⁻¹.
  • Available in small-scale packages, including 25 mg, and as a 10 mM solution in DMSO.
  • Supplied as a hydrochloride salt in solid form for convenient handling.
  • Used to study PDE4 signaling and GABAA receptor modulation in pharmacology and neuroscience research.
Catalog No. 50-002-13330