Learn More
Medchemexpress LLC ETC-159 | 1638250-96-0 | 99.1% | 391.38 | 1 ML
SDP
Catalog No. 5000385221
Encompass_Preferred
ETC-159 is a potent, orally available PORCN inhibitor that inhibits β-catenin reporter activity with an IC50 of 2.9 nM. It is intended for research use only.
  • Inhibits β-catenin reporter activity
  • Potent and orally available PORCN inhibitor
  • Active in multiple cancer models with high Wnt signaling
  • Efficacious in colorectal cancers with R-spondin translocations
  • Good oral pharmacokinetics in mice with 100% oral bioavailability
Catalog No. 50-003-85221 Supplier Medchemexpress LLC Supplier No. HY1898810MM/1ML
ETC-159 is a potent, orally available PORCN inhibitor that inhibits β-catenin reporter activity with an IC50 of 2.9 nM. It is intended for research use only.
  • Inhibits β-catenin reporter activity
  • Potent and orally available PORCN inhibitor
  • Active in multiple cancer models with high Wnt signaling
  • Efficacious in colorectal cancers with R-spondin translocations
  • Good oral pharmacokinetics in mice with 100% oral bioavailability